missing translation for 'onlineSavingsMsg'
Learn More

Methyl perfluorododecanoate, 96%, Thermo Scientific™

Product Code. 11393756
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393756 25 g 25g
11383756 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393756 Supplier Thermo Scientific Alfa Aesar Supplier No. L16809.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56554-52-0
Molecular Formula C13H3F23O2
Molecular Weight (g/mol) 628.128
MDL Number MFCD00236616
InChI Key ABNZIVVYHQYSLI-UHFFFAOYSA-N
Synonym methyl perfluorododecanoate, methyl tricosafluorododecanoate, tricosafluorododecanoic acid methyl ester, dodecanoic acid, tricosafluoro-, methyl ester, methyl tricosafluorolaurate, acmc-1b030, perfluorododecanoic acid methyl ester, methyl tricosafluorolaurate; perfluorododecanoic acid methyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid methyl ester
PubChem CID 547885
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoate
SMILES COC(=O)C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 54°C to 56°C
Density 1.66
Linear Formula CF3(CF2)10CO2CH3
Quantity 25 g
Formula Weight 628.13
Percent Purity 96%
Chemical Name or Material Methyl perfluorododecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.