CAS RN 56643-85-7
IUPAC Name:
4-[(1H-imidazol-1-yl)methyl]aniline
Synonyms:
4-(imidazol-1-ylmethyl)aniline
Molecular Weight (g/mol):
173.22
Molecular Formula:
C10H11N3
InChi Key:
DGHAOTHIDTUSJY-UHFFFAOYSA-N
SMILES:
NC1=CC=C(CN2C=CN=C2)C=C1
1
–
1
of
1
results
1
4-(1h-imidazol-1-ylmethyl)aniline, Thermo Scientific™
CAS: 56643-85-7 Molecular Formula: C10H11N3 Molecular Weight (g/mol): 173.22 MDL Number: MFCD06660785 InChI Key: DGHAOTHIDTUSJY-UHFFFAOYSA-N Synonym: 4-imidazol-1-ylmethylphenylamine,4-1h-imidazol-1-ylmethyl aniline,4-imidazol-1-ylmethyl aniline,4-1h-imidazol-1-yl methyl aniline,4-1-imidazolyl methyl aniline,4-imidazol-1-ylmethyl-phenylamine,1-4-aminobenzyl imidazole,4-imidazolylmethyl phenylamine,1-4-aminobenzyl-1h-imidazole PubChem CID: 1265319 IUPAC Name: 4-(imidazol-1-ylmethyl)aniline SMILES: NC1=CC=C(CN2C=CN=C2)C=C1