missing translation for 'onlineSavingsMsg'
Learn More

4-(1h-imidazol-1-ylmethyl)aniline, Thermo Scientific™

Product Code. 10765373
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10765373 1 g 1g
10236273 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10765373 Supplier Thermo Scientific Maybridge Supplier No. CC43314DA
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 56643-85-7
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.22
MDL Number MFCD06660785
InChI Key DGHAOTHIDTUSJY-UHFFFAOYSA-N
Synonym 4-imidazol-1-ylmethylphenylamine, 4-1h-imidazol-1-ylmethyl aniline, 4-imidazol-1-ylmethyl aniline, 4-1h-imidazol-1-yl methyl aniline, 4-1-imidazolyl methyl aniline, 4-imidazol-1-ylmethyl-phenylamine, 1-4-aminobenzyl imidazole, 4-imidazolylmethyl phenylamine, 1-4-aminobenzyl-1h-imidazole
PubChem CID 1265319
IUPAC Name 4-(imidazol-1-ylmethyl)aniline
SMILES NC1=CC=C(CN2C=CN=C2)C=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 4-(1H-Imidazol-1-ylmethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.