accessibility menu, dialog, popup

UserName

HClH2NN

CAS RN 5705-15-7

IUPAC Name: hydrogen 1-benzyl-1-phenylhydrazine chloride
Synonyms: hydrogen 1-benzyl-1-phenylhydrazine chloride
Molecular Weight (g/mol): 234.73
Molecular Formula: C13H15ClN2
InChi Key: JTYLHYOCBGPMNO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NN(CC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results