CAS RN 57227-09-5
CAS RN 57227-09-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Benzoyl-DL-tyrosyl-N’,N’-dipropylamide, TRC
CAS: 57227-09-5 Molecular Formula: C22 H28 N2 O3 Molecular Weight (g/mol): 368.47 Synonym: (±)-α-(Benzoylamino)-4-hydroxy-N,N-dipropylbenzenepropanamide,CR 1936,N-Benzoyl-DL-tyrosil-N’,N’-dipropylamide,Tiropramide Impurity C; IUPAC Name: N-[1-(dipropylamino)-3-(4-hydroxyphenyl)-1-oxopropan-2-yl]benzamide SMILES: CCCN(CCC)C(=O)C(Cc1ccc(O)cc1)NC(=O)c2ccccc2