accessibility menu, dialog, popup

UserName

CAS RN 57260-71-6

CH3H3CCH3OONNH2

CAS RN 57260-71-6

IUPAC Name: 4-[(tert-butoxy)carbonyl]piperazin-1-ium
Synonyms: 4-(tert-butoxycarbonyl)piperazin-1-ium
Molecular Weight (g/mol): 187.26
Molecular Formula: C9H19N2O2
InChi Key: CWXPZXBSDSIRCS-UHFFFAOYSA-O
SMILES: CC(C)(C)OC(=O)N1CC[NH2+]CC1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (3)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (6)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)