missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl tetrahydropyrazine-1(2H)-carboxylate, 97%, Maybridge

Product Code. 10442303
Click to view available options
:
1g
This item is not returnable. View return policy

Product Code. 10442303

Brand: Thermo Scientific Maybridge AW01051DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 57260-71-6
Assay Percent Range 97%
Molecular Formula C9H19N2O2
MDL Number MFCD00075265
Quantity 1 g
Synonym 1-boc-piperazine, n-boc-piperazine, 1-n-boc-piperazine, tert-butyl 1-piperazinecarboxylate, 1-tert-butoxycarbonyl piperazine, n-t-butoxycarbonylpiperazine, boc-piperazine, n-tert-butoxycarbonylpiperazine, piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 1,1-dimethylethyl ester
InChI Key CWXPZXBSDSIRCS-UHFFFAOYSA-O
SMILES CC(C)(C)OC(=O)N1CC[NH2+]CC1
IUPAC Name 4-[(tert-butoxy)carbonyl]piperazin-1-ium
Molecular Weight (g/mol) 187.26
PubChem CID 143452
Formula Weight 186.25
Chemical Name or Material tert-Butyl tetrahydropyrazine-1(2H)-carboxylate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.