accessibility menu, dialog, popup

UserName

CAS RN 57260-73-8

CH3CH3H3COONHH2N

CAS RN 57260-73-8

IUPAC Name: tert-butyl N-(2-aminoethyl)carbamate
Synonyms: tert-butyl N-(2-aminoethyl)carbamate
Molecular Weight (g/mol): 160.22
Molecular Formula: C7H16N2O2
InChi Key: AOCSUUGBCMTKJH-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCN
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)