missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress NH2-C2-NH-Boc

Product Code. 16407826
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16407826

Brand: MedChemExpress HY40171

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NH2-C2-NH-Boc (PROTAC Linker 22) is a alkyl chain-based PROTAC linker can be used in the synthesis of PROTACs.

Specifications

Purity 97.0%
CAS 57260-73-8
Content And Storage 4°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 160.21 g/mol
Product Type Linker
Quantity 100 mg
Shipping Condition Room temperature
Synonym PROTAC Linker 22
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.