CAS RN 57265-65-3
IUPAC Name:
1-[bis(4-chlorophenyl)methyl]-3-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-2,3-dihydro-1H-imidazole hydrochloridyl
Synonyms:
1-[bis(4-chlorophenyl)methyl]-3-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-2H-imidazole hydrochloridyl
Molecular Weight (g/mol):
688.7
Molecular Formula:
C31H24Cl7N2O
InChi Key:
LBFGPQFQXRJXDI-UHFFFAOYNA-N
SMILES:
[Cl].ClC1=CC=C(C=C1)C(N1CN(CC(OCC2=CC=C(Cl)C=C2Cl)C2=CC=C(Cl)C=C2Cl)C=C1)C1=CC=C(Cl)C=C1
1
–
3
of
3
results
3
Calmidazolium chloride, MedChemExpress
MedChemExpress Calmidazolium chloride (R 24571) is a calmodulin (CaMK) antagonist, antagonizing CaM-dependent phosphodiesterase and calmodulin-induced activation of erythrocyte Ca2+-transporting ATPase with IC50s of 0.15 and 0.35 μM, respectively. Also in anti-cancer research. Calmidazolium binds to CaMK with a Kd of 3 nM.