missing translation for 'onlineSavingsMsg'
Learn More

Calmidazolium chloride, MedChemExpress

Product Code. 16453828
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16453828 10 mg 10mg
16416557 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16453828 Supplier Medchem Express Supplier No. HY103319
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Calmidazolium chloride (R 24571) is a calmodulin (CaMK) antagonist, antagonizing CaM-dependent phosphodiesterase and calmodulin-induced activation of erythrocyte Ca2+-transporting ATPase with IC50s of 0.15 and 0.35 μM, respectively. Also in anti-cancer research. Calmidazolium binds to CaMK with a Kd of 3 nM.

Calmidazolium chloride (R 24571) is a calmodulin (CaMK) antagonist, antagonizing CaM-dependent phosphodiesterase and calmodulin-induced activation of erythrocyte Ca2+-transporting ATPase with IC50s of 0.15 and 0.35 μM, respectively. Also in anti-cancer research. Calmidazolium binds to CaMK with a Kd of 3 nM.

Specifications

Chemical Name or Material Calmidazolium chloride
CAS 57265-65-3
Color White
Purity Grade Notes Research
Molecular Formula C31H23Cl7N2O
Quantity 10 mg
Synonym R 24571
Solubility Information DMSO : 100 mg/mL (145.41 mM; Need ultrasonic)
SMILES ClC1=CC=C(COC(C2=CC=C(Cl)C=C2Cl)CN3C=C[N+](C(C4=CC=C(Cl)C=C4)C5=CC=C(Cl)C=C5)=C3)C(Cl)=C1.[Cl-]
Molecular Weight (g/mol) 687.7
Formula Weight 687.7
Percent Purity 98.93%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.