accessibility menu, dialog, popup

UserName

CAS RN 57729-79-0

IUPAC Name: 2-chloro-6-nitro-4-(trifluoromethyl)aniline
Synonyms: 2-chloro-6-nitro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 240.57
Molecular Formula: C7H4ClF3N2O2
InChi Key: JLWRJMVXRUKFPA-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(C=C1[N+]([O-])=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results