accessibility menu, dialog, popup

UserName

Br

CAS RN 580-13-2

IUPAC Name: 2-bromonaphthalene
Synonyms: 2-bromonaphthalene
Molecular Weight (g/mol): 207.07
Molecular Formula: C10H7Br
InChi Key: APSMUYYLXZULMS-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)