missing translation for 'onlineSavingsMsg'
Learn More

2-Bromonaphthalene, 99%

Product Code. 11944521 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11944521 100 g Glass bottle 100g
10762221 5 g Glass bottle 5g
10052400 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11944521 Supplier Thermo Scientific Acros Supplier No. 179711000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 580-13-2 | C10H7Br | 207.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 580-13-2
Molecular Formula C10H7Br
Molecular Weight (g/mol) 207.07
MDL Number MFCD00004051
InChI Key APSMUYYLXZULMS-UHFFFAOYSA-N
PubChem CID 11372
IUPAC Name 2-bromonaphthalene
SMILES BrC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 53.0°C to 57.0°C
Color Brown-Orange to Yellow
Boiling Point 281.0°C to 282.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C10H7Br
Quantity 100 g
Beilstein 05,548
Merck Index 15,1437
Synonym naphthalene, 2-bromo, 2-naphthyl bromide, 2-bromo-naphthalene, beta-bromonaphthalene, beta-naphthyl bromide, .beta.-bromonaphthalene, unii-syu33i753n, naphthalene,2-bromo, .beta.-naphthyl bromide, pbn-2
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in 8 parts alcohol,ether,chloroform and,benzene
Formula Weight 207.07
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromonaphthalene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.