accessibility menu, dialog, popup

UserName

CAS RN 58356-65-3

CH3OHONNNNMn

CAS RN 58356-65-3

IUPAC Name: manganese(2+) 2,7,12,17-tetraphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide acetic acid
Synonyms: manganese(2+) 2,7,12,17-tetraphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide acetic acid
Molecular Weight (g/mol): 727.73
Molecular Formula: C46H32MnN4O2
InChi Key: UFMFSWMECMUSKE-UHFFFAOYSA-N
SMILES: [Mn++].CC(O)=O.[N-]1C2=CC=C1C(=C1C=CC(=N1)C(=C1[N-]C(C=C1)=C(C1=NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results