missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) acetate meso-tetraphenylporphine, Thermo Scientific™

Product Code. 11393718
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11393718 250 mg 250mg
11383718 1 g 1g
2 options

Product Code. 11393718

Brand: Thermo Scientific Alfa Aesar 039308.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Chemical Identifiers

CAS 58356-65-3
Molecular Formula C46H32MnN4O2
Molecular Weight (g/mol) 727.726
MDL Number MFCD00216548
InChI Key UFMFSWMECMUSKE-UHFFFAOYSA-N
Synonym manganese iii acetate meso-tetraphenylporphine, manganese meso-tetraphenylporphine acetate, manganese iii meso-tetraphenylporphine acetate, 2,7,12,17-tetraphenyl-21,23,24,25-tetraaza-22-manganahexacyclo 9.9.3.1^ 3,6 .1^ 13,16 .0^ 8,23 .0^ 18,21 pentacosa-1,3 25 ,4,6,8,10,12,14,16 24 ,17,19-undecaene; acetic acid
PubChem CID 44135862
IUPAC Name acetic acid;manganese(2+);5,10,15,20-tetraphenylporphyrin-22,24-diide
SMILES CC(=O)O.C1=CC=C(C=C1)C2=C3C=CC(=C(C4=NC(=C(C5=CC=C([N-]5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C=C4)C9=CC=CC=C9)[N-]3.[Mn+2]

Specifications

Quantity 250 mg
Formula Weight 726.71
Physical Form Powder
Chemical Name or Material Manganese(III) acetate meso-tetraphenylporphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.