CAS RN 5840-01-7
IUPAC Name:
1-azatricyclo[6.3.1.0⁴,¹²]dodeca-2,4,6,8(12)-tetraene
Synonyms:
1-azatricyclo[6.3.1.0⁴,¹²]dodeca-2,4,6,8(12)-tetraene
Molecular Weight (g/mol):
157.22
Molecular Formula:
C11H11N
InChi Key:
QCCKSFHMARIKSK-UHFFFAOYSA-N
SMILES:
C1CN2C=CC3=CC=CC(C1)=C23
1
–
1
of
1
results
1
Lilolidine, 98%, Thermo Scientific™
CAS: 5840-01-7 Molecular Formula: C11H11N Molecular Weight (g/mol): 157.21 InChI Key: QCCKSFHMARIKSK-UHFFFAOYSA-N