accessibility menu, dialog, popup

UserName

CAS RN 58479-61-1

IUPAC Name: tert-butyl(chloro)diphenylsilane
Synonyms: tert-butyl(chloro)diphenylsilane
Molecular Weight (g/mol): 274.86
Molecular Formula: C16H19ClSi
InChi Key: MHYGQXWCZAYSLJ-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (4)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (7)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (4)