missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylchlorodiphenylsilane, 98%, AcroSeal™

Product Code. 10778278 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10778278 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10778278 Supplier Thermo Scientific Acros Supplier No. 430921000
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58479-61-1 | C16H19ClSi | 274.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 58479-61-1
Color Brown or Colorless to Yellow
Density 1.0600g/mL
Boiling Point 135.0°C (0.7 mmHg)
Flash Point 149°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Molecular Formula C16H19ClSi
Linear Formula (CH3)3CSi(C6H5)2Cl
Refractive Index 1.5665 to 1.5685
MDL Number MFCD00000497
Quantity 100 mL
Merck Index 15,1568
Specific Gravity 1.06
Synonym tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in non-protic solvents,e.g. aliphatic,or aromatic (chlorinated) hydrocarbons
InChI Key MHYGQXWCZAYSLJ-UHFFFAOYSA-N
SMILES CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
IUPAC Name tert-butyl-chloro-diphenylsilane
Molecular Weight (g/mol) 274.86
PubChem CID 94078
Viscosity 16 mPa.s (25°C)
Formula Weight 274.86
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material tert-Butylchlorodiphenylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butylchlorodiphenylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.