accessibility menu, dialog, popup

UserName

CH3H3CCH3NH3CH3CCH3

CAS RN 585-48-8

IUPAC Name: 2,6-di-tert-butylpyridine
Synonyms: 2,6-di-tert-butylpyridine 2,6-di-T-butylpyridine
Molecular Weight (g/mol): 191.32
Molecular Formula: C13H21N
InChi Key: UWKQJZCTQGMHKD-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=CC(=N1)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (4)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)