missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butylpyridine, 97%

Product Code. 10661063 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10661063 5 g Glass bottle 5g
10333862 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10661063 Supplier Thermo Scientific Acros Supplier No. 204640050
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 585-48-8 | C13H21N | 191.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-48-8
Molecular Formula C13H21N
Molecular Weight (g/mol) 191.32
MDL Number MFCD00006306
InChI Key UWKQJZCTQGMHKD-UHFFFAOYSA-N
Synonym 2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119
PubChem CID 68510
IUPAC Name 2,6-ditert-butylpyridine
SMILES CC(C)(C)C1=NC(=CC=C1)C(C)(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 2.0°C to 4.0°C
Color Yellow
Density 0.8520g/mL
Boiling Point 208.0°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N[C(CH3)3]2
Refractive Index 1.4720 to 1.4740
Quantity 5 g
Beilstein 20, III, 2868
Fieser 01,212
Merck Index 15, 3045
Specific Gravity 0.852
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Formula Weight 191.32
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 6-Di-tert-butylpyridine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Di-tert-butylpyridine
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.