accessibility menu, dialog, popup

UserName

ONOBr

CAS RN 586-78-7

IUPAC Name: 1-bromo-4-nitrobenzene
Synonyms: 4-bromo-1-nitrobenzene
Molecular Weight (g/mol): 202.01
Molecular Formula: C6H4BrNO2
InChi Key: ZDFBKZUDCQQKAC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
Boiling Point 
  • (4)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (4)
  • (3)
  • (3)