accessibility menu, dialog, popup

UserName

BrO

CAS RN 589-10-6

IUPAC Name: (2-bromoethoxy)benzene
Synonyms: benzene, (2-bromoethoxy)-
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: JJFOBACUIRKUPN-UHFFFAOYSA-N
SMILES: BrCCOC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)