missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethyl bromide, 98%

Product Code. 11464547
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464547 500 g 500g
11444547 25 g 25g
11454547 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464547 Supplier Thermo Scientific Alfa Aesar Supplier No. B21087.36
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 589-10-6 | C8H9BrO | 201.063 g/mol

It is used in the synthesis and antihypertensive activity of 4'-substituted spiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-ones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis and antihypertensive activity of 4′-substituted spiro[4H-3,1-benzoxazine-4,4′-piperidin]-2(1H)-ones.

Solubility
Insoluble in water. Solubility in methanol is almost transparent. Soluble in chloroform and ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-10-6
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00000234
InChI Key JJFOBACUIRKUPN-UHFFFAOYSA-N
Synonym 2-bromoethoxy benzene, 2-bromoethyl phenyl ether, beta-bromophenetole, 2-phenoxyethylbromide, 2-phenoxyethyl bromide, 1-bromo-2-phenoxyethane, benzene, 2-bromoethoxy, beta-phenoxyethyl bromide, 2-bromoethylphenylether, 1-2-bromoethoxy benzene
PubChem CID 68526
IUPAC Name 2-bromoethoxybenzene
SMILES C1=CC=C(C=C1)OCCBr

Specifications

Melting Point 31°C to 35°C
Density 1.35
Boiling Point 135°C to 137°C (20 mmHg)
Flash Point 65°C (149°F)
Quantity 500 g
Beilstein 508290
Solubility Information Insoluble in water. Solubility in methanol is almost transparent. Soluble in chloroform and ethyl acetate.
Formula Weight 201.07
Percent Purity 98%
Chemical Name or Material 2-Phenoxyethyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.