accessibility menu, dialog, popup

UserName

OOOOH2OH2OH2OH2OCu

CAS RN 5893-61-8

IUPAC Name: copper(2+) diformate tetrahydrate
Synonyms: copper(2+) diformate tetrahydrate
Molecular Weight (g/mol): 225.64
Molecular Formula: C2H10CuO8
InChi Key: LSIWWRSSSOYIMS-UHFFFAOYSA-L
SMILES: O.O.O.O.[Cu++].[O-]C=O.[O-]C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results