missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) formate tetrahydrate, 98%

Product Code. 11478497
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478497 250 g 250g
11468497 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478497 Supplier Thermo Scientific Alfa Aesar Supplier No. A18569.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5893-61-8 | C2H10CuO8 | 225.64 g/mol

It is employed as an antibacterial agent in the treatment of cellulose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an antibacterial agent in the treatment of cellulose.

Solubility
Soluble in water (125 g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5893-61-8
Molecular Formula C2H10CuO8
Molecular Weight (g/mol) 225.64
MDL Number MFCD00167128
InChI Key LSIWWRSSSOYIMS-UHFFFAOYSA-L
Synonym copper ii formate tetrahydrate, cupric formate tetrahydrate, copper diformate tetrahydrate, unii-v7ra28204j, copper 2+ formate tetrahydrate, cupric formate tetrahydrate mi, copper formate cu hcoo 2 tetrahydrate, formic acid copper salt 2:1 tetrahydrate, acmc-209m8c
PubChem CID 6432244
IUPAC Name copper;diformate;tetrahydrate
SMILES C(=O)[O-].C(=O)[O-].O.O.O.O.[Cu+2]

Specifications

Quantity 250 g
UN Number UN3077
Merck Index 14,2639
Solubility Information Soluble in water (125g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents.
Formula Weight 225.65 (153.59 Anhydrous)
Percent Purity 98%
Chemical Name or Material Copper(II) formate tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.