accessibility menu, dialog, popup

UserName

OHHSHS

CAS RN 59-52-9

IUPAC Name: 2,3-disulfanylpropan-1-ol
Synonyms: dimercaprol bal in oil sulfactin british anti-lewisite dimercaprolum british anti-lewisite agent antoxol british anti lewisite agent dimersol BAL
Molecular Weight (g/mol): 124.22
Molecular Formula: C3H8OS2
InChi Key: WQABCVAJNWAXTE-UHFFFAOYNA-N
SMILES: OCC(S)CS
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)