missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimercaptopropanol, 97%

Product Code. 11396726
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11396726

Brand: Thermo Scientific Alfa Aesar L03953.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59-52-9 | C3H8OS2 | 124.216 g/mol

British Anti-Lewisite. A chelator that inhibits sugar nucleotide degradation.2,3-Dimercaptopropanol is used as a chelating agent involved in the treatment of arsenic, antimony, lead, gold and mercury poisoning. It is also used as an antidote to the warfare agent Lewisite. Further, it is used for the treatment of Wilson's disease. In addition to this, it is used as an adjunct in the treatment of the acute encephalopathy of lead toxicity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water, ether, ethanol, vegetable oils and acetone. Slightly miscible with chloroform.

Notes
Air sensitive. Store in a cool place. Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-52-9
Molecular Formula C3H8OS2
Molecular Weight (g/mol) 124.216
MDL Number MFCD00004864
InChI Key WQABCVAJNWAXTE-UHFFFAOYSA-N
Synonym dimercaprol, 2,3-dimercapto-1-propanol, 2,3-dimercaptopropanol, dithioglycerine, dicaptol, sulfactin, dimercaptopropanol, british anti-lewisite, 2,3-dithiopropanol, antoxol
PubChem CID 3080
ChEBI CHEBI:64198
IUPAC Name 2,3-bis(sulfanyl)propan-1-ol
SMILES C(C(CS)S)O

Specifications

Density 1.244
Boiling Point 120°C (15 mmHg)
Flash Point >110°C (230°F)
Odor Garlic-like
Refractive Index 1.574
Quantity 5 g
UN Number UN2810
Beilstein 1732058
Sensitivity Air Sensitive
Merck Index 14,3209
Solubility Information Miscible with water,ether,ethanol,vegetable oils and acetone. Slightly miscible with chloroform.
Formula Weight 124.22
Percent Purity 97%
Chemical Name or Material 2,3-Dimercaptopropanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.