CAS RN 591-12-8
CAS RN 591-12-8
IUPAC Name:
5-methyl-2,3-dihydrofuran-2-one
Synonyms:
5-methyl-2(3H)-furanone delta(2)-angelica lactone α-angelica lactone 5-methylfuranone delta(sup 2)-angelica lactone α-angelicalactone β,gamma-angelica lactone α-angelicalacton
Molecular Weight (g/mol):
98.1
Molecular Formula:
C5H6O2
InChi Key:
QOTQFLOTGBBMEX-UHFFFAOYSA-N
SMILES:
CC1=CCC(=O)O1
1
–
1
of
1
results
1
alpha-Angelicalactone, 98%
CAS: 591-12-8 Molecular Formula: C5H6O2 Molecular Weight (g/mol): 98.10 MDL Number: MFCD00005375 InChI Key: QOTQFLOTGBBMEX-UHFFFAOYSA-N Synonym: alpha-angelica lactone,alpha-angelicalactone,5-methyl-2 3h-furanone,5-methylfuran-2 3h-one,2 3h-furanone, 5-methyl,4-hydroxypent-3-enoic acid lactone,delta 2-angelica lactone,4-hydroxy-3-pentenoic acid gamma-lactone,gamma-methyl-beta,gamma-crotonolactone,.alpha.-angelica lactone PubChem CID: 11559 ChEBI: CHEBI:36433 IUPAC Name: 5-methyl-3H-furan-2-one SMILES: CC1=CCC(=O)O1