missing translation for 'onlineSavingsMsg'
Learn More

alpha-Angelicalactone, 98%

Product Code. 11341977
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341977 100 g 100g
11321977 5 g 5g
11331977 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11341977 Supplier Thermo Scientific Alfa Aesar Supplier No. L07602.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-12-8 | C5H6O2 | 98.10 g/mol

alpha-Angelicalactone acts as acancer chemopreventive agent. It is used in the synthesis of glutathione and the activity of glutathione-S-transferase and UDP-glucunonosyltransferase detoxification enzymes in esophagus, stomach, intestine, and liver. It is used in oral care formulations to round out mint notes and in coffee flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Angelicalactone acts as acancer chemopreventive agent. It is used in the synthesis of glutathione and the activity of glutathione-S-transferase and UDP-glucunonosyltransferase detoxification enzymes in esophagus, stomach, intestine, and liver. It is used in oral care formulations to round out mint notes and in coffee flavors.

Solubility
Soluble in water (5g/100mL) and alcohol.

Notes
Moisture-sensitive. Store under inert gas. Protect from moisture. Incompatible materials are Oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-12-8
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00005375
InChI Key QOTQFLOTGBBMEX-UHFFFAOYSA-N
Synonym alpha-angelica lactone, alpha-angelicalactone, 5-methyl-2 3h-furanone, 5-methylfuran-2 3h-one, 2 3h-furanone, 5-methyl, 4-hydroxypent-3-enoic acid lactone, delta 2-angelica lactone, 4-hydroxy-3-pentenoic acid gamma-lactone, gamma-methyl-beta,gamma-crotonolactone, .alpha.-angelica lactone
PubChem CID 11559
ChEBI CHEBI:36433
IUPAC Name 5-methyl-3H-furan-2-one
SMILES CC1=CCC(=O)O1

Specifications

Melting Point 17°C to 18°C
Density 1.093
Boiling Point 53°C to 54°C (10 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.448
Quantity 100 g
Beilstein 108394
Merck Index 14,647
Solubility Information Soluble in water (5g/100mL) and alcohol.
Formula Weight 98.1
Percent Purity 98%
Chemical Name or Material alpha-Angelicalactone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.