CAS RN 593-49-7
CAS RN 593-49-7
IUPAC Name:
heptacosane
Synonyms:
heptacosane
Molecular Weight (g/mol):
380.75
Molecular Formula:
C27H56
InChi Key:
BJQWYEJQWHSSCJ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCCCCCCCCCC
1
–
3
of
3
results
1
n-Heptacosane, 97%
CAS: 593-49-7 Molecular Formula: C27H56 Molecular Weight (g/mol): 380.745 MDL Number: MFCD00009862 InChI Key: BJQWYEJQWHSSCJ-UHFFFAOYSA-N Synonym: n-heptacosane,unii-vp371w2gjs,vp371w2gjs,hexacosane, methyl,acmc-1aog8,qspl 047,qspl 073,heptacosane, analytical standard,ch3-ch2 25-ch3,heptacosane gc PubChem CID: 11636 ChEBI: CHEBI:32941 IUPAC Name: heptacosane SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCC
2
n-Heptacosane 100 μg/mL in Hexane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality