CAS RN 59303-12-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(Methoxymethyl)-6-methylpyridine, TRC
CAS: 59303-12-7 Molecular Formula: C8H11NO Molecular Weight (g/mol): 137.17 Synonym: NSC 76084 IUPAC Name: 2-(methoxymethyl)-6-methylpyridine SMILES: COCc1cccc(C)n1