missing translation for 'onlineSavingsMsg'
Learn More

2-(Methoxymethyl)-6-methylpyridine, TRC

Product Code. 30479562
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479562 250 mg 250mg
30471900 500 mg 500mg
30475423 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479562 Supplier Toronto Research Chemicals Supplier No. TRCM336793250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59303-12-7
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.17
Synonym NSC 76084
IUPAC Name 2-(methoxymethyl)-6-methylpyridine
SMILES COCc1cccc(C)n1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H11NO/c1-7-4-3-5-8(9-7)6-10-2/h3-5H,6H2,1-2H3
Formula Weight 137.0841
Chemical Name or Material 2-(Methoxymethyl)-6-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.