accessibility menu, dialog, popup

UserName

H2NOClClCl

CAS RN 594-65-0

IUPAC Name: trichloroacetamide
Synonyms: trichloroacetamide
Molecular Weight (g/mol): 162.39
Molecular Formula: C2H2Cl3NO
InChi Key: UPQQXPKAYZYUKO-UHFFFAOYSA-N
SMILES: NC(=O)C(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results