accessibility menu, dialog, popup

UserName

FFFOOOFFFFFFF

CAS RN 59483-84-0

IUPAC Name: bis(2,3,4,5,6-pentafluorophenyl) carbonate
Synonyms: bis(2,3,4,5,6-pentafluorophenyl) carbonate
Molecular Weight (g/mol): 394.12
Molecular Formula: C13F10O3
InChi Key: IOVVFSGCNWQFQT-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)OC2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results