accessibility menu, dialog, popup

UserName

ClH3CONH2H2N

CAS RN 59548-39-9

IUPAC Name: 4-methoxybenzene-1,2-diamine hydrochloridyl
Synonyms: 4-methoxybenzene-1,2-diamine hydrochloridyl
Molecular Weight (g/mol): 173.62
Molecular Formula: C7H10ClN2O
InChi Key: NOFDEJFDTTYTCG-UHFFFAOYSA-N
SMILES: [Cl].COC1=CC=C(N)C(N)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results