accessibility menu, dialog, popup

UserName

OOOO

CAS RN 59725-59-6

IUPAC Name: 3-[2-(benzoyloxy)phenyl]propanoate
Synonyms: 3-[2-(benzoyloxy)phenyl]propanoate
Molecular Weight (g/mol): 269.28
Molecular Formula: C16H13O4
InChi Key: WZLUNIRPOQOEJR-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCC1=CC=CC=C1OC(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results