accessibility menu, dialog, popup

UserName

CH3CH3CH3ON

CAS RN 59997-51-2

IUPAC Name: 4,4-dimethyl-3-oxopentanenitrile
Synonyms: 4,4-dimethyl-3-oxopentanenitrile
Molecular Weight (g/mol): 125.17
Molecular Formula: C7H11NO
InChi Key: MXZMACXOMZKYHJ-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)CC#N
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)