missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetylacetonitrile, 99%, Thermo Scientific Chemicals

Product Code. 11457777
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457777 25 g 25g
11467777 100 g 100g
11477777 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457777 Supplier Thermo Scientific Alfa Aesar Supplier No. A16309.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

Trimethylacetylacetonitrile is a useful intermediate. 4,4-Dimethyl-3-oxopentanenitrile has been used in the preparation of 4,4-dimethyl-3-oxo-2-benzylpentanenitrile, 4,4-dimethyl-3-oxo-2-(4-hydroxybenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(4-methoxybenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(4-fluorobenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(3,4-methylenedioxybenzyl)pentanenitrile, 1-(5-tert-butyl-2-p-tolyl-2H-pyrazol-3-yl)-3-phenylurea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetylacetonitrile is a useful intermediate. 4,4-Dimethyl-3-oxopentanenitrile has been used in the preparation of 4,4-dimethyl-3-oxo-2-benzylpentanenitrile, 4,4-dimethyl-3-oxo-2-(4-hydroxybenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(4-methoxybenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(4-fluorobenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(3,4-methylenedioxybenzyl)pentanenitrile, 1-(5-tert-butyl-2-p-tolyl-2H-pyrazol-3-yl)-3-phenylurea.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59997-51-2
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.171
MDL Number MFCD00010208
InChI Key MXZMACXOMZKYHJ-UHFFFAOYSA-N
Synonym pivaloylacetonitrile, trimethylacetylacetonitrile, 4,4-dimethyl-3-oxovaleronitrile, pentanenitrile, 4,4-dimethyl-3-oxo, 4,4-dimethyl-3-oxo-pentanenitrile, cyanopinacolone, pivaloyl acetonitrile, 1-cyano-3,3-dimethyl-2-butanone, valeronitrile, 4,4-dimethyl-3-oxo, pyvaolylacetonitrile
PubChem CID 108871
IUPAC Name 4,4-dimethyl-3-oxopentanenitrile
SMILES CC(C)(C)C(=O)CC#N

Specifications

Melting Point 66°C to 70°C
Quantity 25 g
UN Number UN3439
Beilstein 1746674
Solubility Information Insoluble in water.
Formula Weight 125.17
Percent Purity 99%
Chemical Name or Material Trimethylacetylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.