accessibility menu, dialog, popup

UserName

CAS RN 60-35-5

IUPAC Name: acetamide
Synonyms: acetamide azetamid essigsaeureamid ethanamid
Molecular Weight (g/mol): 59.07
Molecular Formula: C2H5NO
InChi Key: DLFVBJFMPXGRIB-UHFFFAOYSA-N
SMILES: CC(N)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (3)
  • (1)
Grade 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (5)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)