missing translation for 'onlineSavingsMsg'
Learn More

Acetamide, Extra Pure, SLR, Fisher Chemical

Product Code. 10385640 Shop All Fisher Chemical Products
Click to view available options
:
100g, Plastic powder jar
This item is not returnable. View return policy

Product Code. 10385640

Brand: Fisher Chemical A/0160/48

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 60-35-5
Melting Point 79°C
Color White
Boiling Point 221°C
Packaging Plastic powder jar
Molecular Formula C2H5NO
MDL Number 8023
Quantity 100 g
Formula Weight 59.06g/mol
Physical Form Solid
compliance-icons
Product Identifier
  • Acetamide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.