accessibility menu, dialog, popup

UserName

H3COOCH3CH3Br

CAS RN 600-00-0

IUPAC Name: ethyl 2-bromo-2-methylpropanoate
Synonyms: ethyl α-bromoisobutyrate
Molecular Weight (g/mol): 195.06
Molecular Formula: C6H11BrO2
InChi Key: IOLQWGVDEFWYNP-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)(C)Br
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)