missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromoisobutyrate, 98+%

Product Code. 11419827
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419827 250 g 250g
11409827 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419827 Supplier Thermo Scientific Alfa Aesar Supplier No. B22785.30
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 600-00-0 | C6H11BrO2 | 195.06 g/mol

Ethyl 2-bromoisobutyrate is used as an Atom Transfer Radical Polymerization (ATRP) initiator that will generate a polymer with an ethyl end group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-bromoisobutyrate is used as an Atom Transfer Radical Polymerization (ATRP) initiator that will generate a polymer with an ethyl end group.

Solubility
Soluble in alcohol, ether. Insoluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents, acids, bases, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 600-00-0
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000123
InChI Key IOLQWGVDEFWYNP-UHFFFAOYSA-N
Synonym ethyl 2-bromoisobutyrate, ethyl 2-bromo-2-methylpropionate, ethyl alpha-bromoisobutyrate, ethyl .alpha.-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, ethyl ester, 2-bromo-2-methylpropanoic acid, ethyl ester, ethyl-2-bromoisobutyrate, propanoic acid, bromo-2-methyl-, ethyl ester, ethyl 2-bromo-2-methyl-propanoate, propionic acid, 2-bromo-2-methyl-, ethyl ester
PubChem CID 11745
IUPAC Name ethyl 2-bromo-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)Br

Specifications

Density 1.329
Boiling Point 162°C to 164°C
Flash Point 57°C (135°F)
Refractive Index 1.444
Quantity 250 g
UN Number UN2924
Beilstein 1098947
Solubility Information Soluble in alcohol,ether. Insoluble in water.
Formula Weight 195.06
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-bromoisobutyrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.