accessibility menu, dialog, popup

UserName

HOOHONOO

CAS RN 602-00-6

IUPAC Name: 3-hydroxy-2-nitrobenzoic acid
Synonyms: 3-hydroxy-2-nitrobenzoic acid
Molecular Weight (g/mol): 183.12
Molecular Formula: C7H5NO5
InChi Key: KPDBKQKRDJPBRM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(O)=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results