accessibility menu, dialog, popup

UserName

CAS RN 603-80-5

CH3OHOO

CAS RN 603-80-5

IUPAC Name: 3-hydroxy-2-methylbenzoate
Synonyms: 3-hydroxy-2-methylbenzoate
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H7O3
InChi Key: RIERSGULWXEJKL-UHFFFAOYSA-M
SMILES: CC1=C(O)C=CC=C1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results