accessibility menu, dialog, popup

UserName

CH3NH2NOO

CAS RN 603-83-8

IUPAC Name: 2-methyl-3-nitroaniline
Synonyms: 3-nitro-o-toluidine
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: HFCFJYRLBAANKN-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)