accessibility menu, dialog, popup

UserName

OSOONONSClOO(?)

CAS RN 60311-02-6

IUPAC Name: 16-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3λ⁴-oxa-9,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,3,5(28),13,15,17,19(27)-heptaen-3-ylium
Synonyms: sulforhodamine 101
Molecular Weight (g/mol): 625.15
Molecular Formula: C31H29ClN2O6S2
InChi Key: MPLHNVLQVRSVEE-UHFFFAOYSA-N
SMILES: [O-]S(=O)(=O)C1=CC(=CC=C1C1=C2C=C3CCCN4CCCC(C2=[O+]C2=C5CCCN6CCCC(C=C12)=C56)=C34)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results