accessibility menu, dialog, popup

UserName

CH3CH3HO

CAS RN 6032-29-7

IUPAC Name: pentan-2-ol
Synonyms: 2-pentanol
Molecular Weight (g/mol): 88.15
Molecular Formula: C5H12O
InChi Key: JYVLIDXNZAXMDK-UHFFFAOYNA-N
SMILES: CCCC(C)O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (6)
  • (3)
  • (2)
  • (1)