accessibility menu, dialog, popup

UserName

CAS RN 60367-52-4

NHNClO

CAS RN 60367-52-4

IUPAC Name: 5-chloro-2-phenyl-1H-imidazole-4-carbaldehyde
Synonyms: 5-chloro-2-phenyl-1H-imidazole-4-carbaldehyde
Molecular Weight (g/mol): 206.63
Molecular Formula: C10H7ClN2O
InChi Key: CCXUATWKEDQRMP-UHFFFAOYSA-N
SMILES: ClC1=C(C=O)N=C(N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results