accessibility menu, dialog, popup

UserName

CAS RN 60480-83-3

CH3HNNH2CH3HCl

CAS RN 60480-83-3

IUPAC Name: (2,4-dimethylphenyl)hydrazine hydrochloride
Synonyms: (2,4-dimethylphenyl)hydrazine hydrochloride
Molecular Weight (g/mol): 172.66
Molecular Formula: C8H13ClN2
InChi Key: ZLGXGLXFBMIVPU-UHFFFAOYSA-N
SMILES: Cl.CC1=CC(C)=C(NN)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results